Learn More
STEMCELL Technologies Anti-Mouse EPCR Antibody, Clone RMEPCR1560 (1560), FITC, Size: 25 Tests
SDP
Supplier:  STEMCELL Technologies 60038FI.1
Encompass
Clone RMEPCR1560 (1560) is a rat monoclonal antibody that reacts with mouse EPCR (CD201), an ~25 kDa type I transmembrane glycoprotein expressed by endothelial cells, subsets of hematopoietic stem cells (HSCs) and dendritic cells, and several malignant cell lines. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of EPCR (CD201). EPCR exhibits homology with the MHC class 1/CD1 protein family. EPCR binds protein C and activated protein C, thus augmenting protein C activation by the thrombin-thrombomodulin complex and regulating blood coagulation and inflammation. EPCR expression is associated with the stem cell activity of bone marrow cell populations isolated using conventional markers, indicating the usefulness of EPCR as a single marker for the identification of mouse HSCs. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5249