Learn More
Medchemexpress LLC Anti-Mouse F4 80 Ant 1mg | 1mg
SDP
Catalog No. 5000324881
Encompass_Preferred
Anti-Mouse F4/80 Antibody (CI A3-1) is an anti-mouse F4/80 IgG2b monoclonal antibody Anti-Mouse F4/80 Antibody (CI A3-1) can clear macrophages Anti-Mouse F4/80 Antibody (CI A3-1) can be used for research on inflammation conditions such as endometriosis Anti-Mouse F4/80 Antibody (CI A3-1) often used for flow cytometry analysis and immunohistochemical analysis[1][2]
Catalog No. 50-003-24881 Supplier Medchemexpress LLC Supplier No. HYP9902461MG
Anti-Mouse F4/80 Antibody (CI A3-1) is an anti-mouse F4/80 IgG2b monoclonal antibody Anti-Mouse F4/80 Antibody (CI A3-1) can clear macrophages Anti-Mouse F4/80 Antibody (CI A3-1) can be used for research on inflammation conditions such as endometriosis Anti-Mouse F4/80 Antibody (CI A3-1) often used for flow cytometry analysis and immunohistochemical analysis[1][2]