Learn More
STEMCELL Technologies Anti-Mouse F4/80 Antibody, Clone BM8, PE, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60027PE
Encompass
Clone BM8 is a rat monoclonal antibody that reacts with the F4/80 antigen (also termed Ly-71 in mouse and EMR-1 in human), an ~160 kDa transmembrane glycoprotein belonging to the EGF-TM7 family of G-protein-coupled receptors. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of human and mouse F4/80. F4/80 is reportedly the only macrophage marker suitable for distinguishing destructive from non-destructive inflammatory processes in the pancreas. The protein is thought to play a role in the generation of CD8+ regulatory T cells involved in immune tolerance. This antibody clone has also been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Monocyte Enrichment Kit (Catalog #19761) and EasySep™ Mouse CD11b Positive Selection Kit (Catalog #18770). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5136