Learn More
Bio X Cell Anti-Mouse GITR, Clone DTA-1, IgG2b, 1 MG
Supplier: Bio X Cell BE0063R001MG
The DTA-1 monoclonal antibody targets mouse GITR, a 66-70 kDa co-stimulatory immune checkpoint molecule of the Tumor Necrosis Factor superfamily. This agonistic antibody is commonly used to induce GITR signaling in vivo.
- Reacts with mouse GITR (glucocorticoid-induced TNFR-related gene)
- Clone DTA-1, an agonistic antibody
- Isotype rat IgG2b, λ
- Useful for in vivo GITR stimulation
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Protein G purified from animal-free cell culture supernatant
- Purity ≥95% by SDS-PAGE
- Endotoxin ≤1 EU/mg (≤0.001 EU/μg)
- Sterile filtered via 0.2 μm filtration
- Molecular weight 150 kDa
- Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.