Learn More
Bio X Cell RecombiMAb Anti-Mouse GITR DTA-1-CP029 Clone, 1 MG
Supplier: Bio X Cell CP029R001MG
The recombinant DTA-1-CP029 monoclonal antibody is a chimeric version of the original DTA-1, engineered with LALA-PG mutations to prevent binding to murine Fcγ receptors. This modification reduces anaphylaxis in mice while preserving T cell agonistic and antitumor efficacy. It targets mouse GITR, a co-stimulatory immune checkpoint molecule on T lymphocytes and regulatory T cells.
- Clone: DTA-1-CP029
- Host: Mouse
- Target: Mouse GITR
- Isotype: Mouse IgG2a LALA-PG, κ
- Purity: ≥95% by SDS-PAGE
- Endotoxin: ≤0.5EU/mg
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Storage: 4°C; do not freeze
- Applications: Flow cytometry, Western Blot, Immunoprecipitation, in vitro/in vivo T cell stimulation, in vivo antitumor immunity
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.