Learn More
STEMCELL Technologies Anti-Mouse Gr-1 Antibody, Clone RB6-8C5, FITC, Size: 500 µg
SDP
Supplier:  STEMCELL Technologies 60028FI
Encompass
Rat monoclonal IgG2b antibody against mouse Gr-1 (Ly-6G/Ly-6C), FITC-conjugated
Catalog No. 50-197-5147