Learn More
Rockland Immunochemicals Mouse κ (kappa chain) Antibody Peroxidase Conjugated 1 mg
SDP
Catalog No. 501943907
Encompass_Preferred
Anti-Mouse κ (kappa chain) (RABBIT) Antibody generated in rabbit detects specifically Mouse kappa light chain. Immunoglobulins are heterotetramers composed of 2 immunoglobulin heavy and 2 immunoglobulin light chains. The immunoglobulin light chain is the small polypeptide subunit of an antibody (immunoglobulin). The light chains can be categorized into kappa type or lambda type and both are used to construct the antigen binding F(ab) region of an antibody along with the variable region of the heavy chain. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition. Anti-Mouse IgG kappa is conjugated to peroxidase.
Catalog No. 50-194-3907 Supplier Rockland Immunochemicals Supplier No. 6104310
Anti-Mouse κ (kappa chain) (RABBIT) Antibody generated in rabbit detects specifically Mouse kappa light chain. Immunoglobulins are heterotetramers composed of 2 immunoglobulin heavy and 2 immunoglobulin light chains. The immunoglobulin light chain is the small polypeptide subunit of an antibody (immunoglobulin). The light chains can be categorized into kappa type or lambda type and both are used to construct the antigen binding F(ab) region of an antibody along with the variable region of the heavy chain. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition. Anti-Mouse IgG kappa is conjugated to peroxidase.