Learn More
Medchemexpress LLC Anti-Mouse Human CD1 5mg | 5mg
SDP
Catalog No. 5000323536
Encompass_Preferred
Anti-Mouse/Human CD11b Antibody (M1/70) is an anti-mouse CD11b IgG2b monoclonal antibody Anti-Mouse/Human CD11b Antibody (M1/70) can significantly inhibit the adhesion between dendritic cells (DCs) and platelets Anti-Mouse/Human CD11b Antibody (M1/70) can kill ovarian cancer cells and inhibit their migration Anti-Mouse/Human CD11b Antibody (M1/70) can alleviate renal fibrosis and inflammation Anti-Mouse/Human CD11b Antibody (M1/70) can be used for researches on inflammation conditions and cancer such as ischemia-reperfusion injury (IRI) thrombotic inflammatory conditions and ovarian cancer[1][2][3]
Catalog No. 50-003-23536 Supplier Medchemexpress LLC Supplier No. HYP991215MG
Anti-Mouse/Human CD11b Antibody (M1/70) is an anti-mouse CD11b IgG2b monoclonal antibody Anti-Mouse/Human CD11b Antibody (M1/70) can significantly inhibit the adhesion between dendritic cells (DCs) and platelets Anti-Mouse/Human CD11b Antibody (M1/70) can kill ovarian cancer cells and inhibit their migration Anti-Mouse/Human CD11b Antibody (M1/70) can alleviate renal fibrosis and inflammation Anti-Mouse/Human CD11b Antibody (M1/70) can be used for researches on inflammation conditions and cancer such as ischemia-reperfusion injury (IRI) thrombotic inflammatory conditions and ovarian cancer[1][2][3]