Learn More
Selleck Chemical LLC Anti-mouse IFN-gamma-InVivo
SDP
Catalog No. 503136568
Encompass_Preferred
The R4-6A2 monoclonal antibody reacts with mouse IFN (interferon gamma) which is a 20 kDa soluble pleiotropic cytokine and the sole member of the type II class of interferons The R4-6A2 antibody has been shown to neutralize both natural and recombinant IFN
Catalog No. 50-313-6568 Supplier Selleck Chemical LLC Supplier No. A21055MG
The R4-6A2 monoclonal antibody reacts with mouse IFN (interferon gamma) which is a 20 kDa soluble pleiotropic cytokine and the sole member of the type II class of interferons The R4-6A2 antibody has been shown to neutralize both natural and recombinant IFN