Learn More
Selleck Chemical LLC Anti-mouse IFNAR-1-InVivo
SDP
Supplier:  Selleck Chemical LLC A21211MG
Encompass_Preferred
Anti-mouse IFNAR-1-InVivo monoclonal antibody (Clone MAR1-5A3) reacts with mouse IFNAR-1 (IFN alpha/beta receptor subunit 1) Anti-mouse IFNAR-1-InVivo has been shown to inhibit Type I IFN receptor signaling in vitro and in vivo
Catalog No. 50-234-0925