Learn More
PBL Assay Science Anti-Mouse Interferon Beta (IFN-Beta) Rat Monoclonal Antibody, Clone RMMB-1 (MAb) [250 micrograms (ug)]
Supplier:  PBL Assay Science 224001
Encompass_Preferred
Rat monoclonal antibody Anti-Mouse Interferon Beta, Clone RMMB-1 is manufactured by PBL Assay Science (Catalog #22400-1) for research use only (RUO) and packaged in 250 micrograms (ug). This MAb binds to mouse IFN beta and weakly neutralizes mouse IFN beta. It is suitable for use in direct binding ELISA applications.
Catalog No. 50-153-8034