Learn More
PBL Assay Science Anti-Mouse Interferon Gamma (IFN-Gamma) Rat Monoclonal Antibody, Clone RMMG-1 (MAb) [500 micrograms (ug)]
Supplier:  PBL Assay Science 225001
Encompass_Preferred
Rat monoclonal antibody Anti-Mouse Interferon Gamma, Clone RMMG-1 is manufactured by PBL Assay Science (Catalog #22500-1) for research use only (RUO) and packaged in 500 micrograms (ug). This MAb binds to mouse IFN gamma with high affinity. It is suitable for use in neutralization, immunohistochemistry, immunoprecipitation, ELISA, and Western Blot applications.
Catalog No. 50-153-8035