Learn More
Medchemexpress LLC Anti-Mouse IFNR CD1 5mg | 5mg
SDP
Catalog No. 5000324882
Encompass_Preferred
Anti-Mouse IFN R/CD119 Antibody (GR-20) is a rat-derived IgG2a type antibody inhibitor targeting to mouse IFN R/CD119 Anti-Mouse IFN R/CD119 Antibody (GR-20) reacts with the mouse IFN R (interferon gamma receptor) chain also known as CD119 and IFN receptor 1 Anti-Mouse IFN R/CD119 Antibody (GR-20) blocks the binding of IFN to CD119 therefore inhibiting IFN signaling Anti-Mouse IFN R/CD119 Antibody (GR-20) can be used for the researches of cancer infection inflammation and immunology such as melanoma and C trachomatis infected genital tract inflammation[1][2]
Catalog No. 50-003-24882 Supplier Medchemexpress LLC Supplier No. HYP9902335MG
Anti-Mouse IFN R/CD119 Antibody (GR-20) is a rat-derived IgG2a type antibody inhibitor targeting to mouse IFN R/CD119 Anti-Mouse IFN R/CD119 Antibody (GR-20) reacts with the mouse IFN R (interferon gamma receptor) chain also known as CD119 and IFN receptor 1 Anti-Mouse IFN R/CD119 Antibody (GR-20) blocks the binding of IFN to CD119 therefore inhibiting IFN signaling Anti-Mouse IFN R/CD119 Antibody (GR-20) can be used for the researches of cancer infection inflammation and immunology such as melanoma and C trachomatis infected genital tract inflammation[1][2]