Learn More
Rockland Immunochemicals Anti-MOUSE IgG F(c) (RABBIT) Antibody - 610-4103
SDP
Supplier:  Rockland Immunochemicals 6104103
Encompass
Anti-Mouse IgG F(c) generated in rabbit is a proteolytic fragment of immunoglobulin G (IgG) obtained by limited digestion with the enzyme papain under controlled conditions of temperature, time and pH. Receptors bind the Fc portion of mouse IgG and often this fragment is removed from immunoglobulins to minimize receptor binding and lower background reactivity.
Catalog No. NC9992488