Learn More
Enzo Life Sciences Goat anti-mouse IgG F(ab')2, polyclonal antibody (HRP conjugate) (1 ml)
SDP
Supplier:  Enzo Life Sciences ADISAB100J
Encompass_Preferred
Host: Goat. Immunogen: Fab fragment of mouse IgG. Species Reactivity: Mouse. Specificity: Based on immunoelectrophoresis and/or ELISA, the antibody reacts with the F(ab’)2/Fab portion of mouse IgG, and also with the light chains of other mouse immunoglobulins. No antibody was detected against the Fc portion of mouse IgG or against other non-immunoglobulin serum proteins. Applications: ELISA, IHC, WB. Recommended dilutions: ELISA (1:2,500-1:50,000). Immunohistochemistry (1:250-1:2500). Western Blot (1:2500-1:50,000 colorimetric, 1:5,000-1:100,000 ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Immunoaffinity purified. Formulation: Liquid. In 0.005M sodium phosphate, pH 7.6, containing 0.125M sodium chloride, 7.5mg/ml BSA (IgG-free, protease-free), and 50% glycerol.
Catalog No. 50-667-383