Learn More
Seracare Life Sciences Inc ANTI-MOUSE IGG (H+L) AB RSA
Supplier:  Seracare Life Sciences Inc 52100191
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgG, both heavy and light chains (whole IgG) made in Goat. Product has been cross-adsorbed with rat immunoglobulin to reduce cross-reactivity. Product is in lyophilized form.
Catalog No. 50-668-29