Learn More
Rockland Immunochemicals Mouse IgG (H&L) Secondary Antibody Alkaline Phosphatase Conjugated 1 mg
SDP
Catalog No. 501943910
Encompass_Preferred
Anti-Mouse IgG Alkaline Phosphatase antibody generated in rabbit detects reactivity to Mouse IgG. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.
Catalog No. 50-194-3910 Supplier Rockland Immunochemicals Supplier No. 6104502
Anti-Mouse IgG Alkaline Phosphatase antibody generated in rabbit detects reactivity to Mouse IgG. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.