Learn More
Rockland Immunochemicals Mouse IgG (H&L) Secondary Antibody Alkaline Phosphatase Conjugated Pre-Adsorbed 1 mg
SDP
Supplier:  Rockland Immunochemicals 610105121
Encompass
Anti-Mouse IgG Alkaline Phosphatase Antibody generated in goat detects reactivity to Mouse IgG. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.
Catalog No. 50-194-3897