Learn More
Rockland Immunochemicals Anti-MOUSE IgG (H&L) (DONKEY) Antibody DyLight™ 680 Conjugated (Min X Bv Ch Gt GP Ham Hs Hu Rb Rt & Sh Serum Proteins) - 610-744-124
SDP
Catalog No. 501058504
Encompass_Preferred
Anti-Mouse IgG DyLight™680 Antibody generated in donkey detects reactivity to Mouse IgG. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Rabbit, Rat and Sheep Serum Proteins.  This antibody will react with heavy chains of mouse IgG and with light chains of most mouse immunoglobulins. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This product is designed for IF, fluorescence based plate assays (FLISA) and fluorescent WB.
Catalog No. 50-105-8504 Supplier Rockland Immunochemicals Supplier No. 610744124
Anti-Mouse IgG DyLight™680 Antibody generated in donkey detects reactivity to Mouse IgG. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Rabbit, Rat and Sheep Serum Proteins.  This antibody will react with heavy chains of mouse IgG and with light chains of most mouse immunoglobulins. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This product is designed for IF, fluorescence based plate assays (FLISA) and fluorescent WB.