Learn More
Rockland Immunochemicals Anti-MOUSE IgG (H&L) (DONKEY) Antibody DyLight™ 680 Conjugated - 610-744-002
SDP
Catalog No. 501058503
Encompass_Preferred
Anti-Mouse IgG DyLight™680 Antibody generated in donkey detects reactivity to Mouse IgG. This antibody will react with heavy chains of Mouse IgG and with light chains of most Mouse immunoglobulins. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms. The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Catalog No. 50-105-8503 Supplier Rockland Immunochemicals Supplier No. 610744002
Anti-Mouse IgG DyLight™680 Antibody generated in donkey detects reactivity to Mouse IgG. This antibody will react with heavy chains of Mouse IgG and with light chains of most Mouse immunoglobulins. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms. The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.