Learn More
Rockland Immunochemicals Anti-MOUSE IgG (H&L) (GOAT) Antibody DyLight™ 680 Conjugated (Min X Bv Ch Gt GP Ham Hs Hu Rb Rt & Sh Serum Proteins) - 610-144-121
SDP
Supplier:  Rockland Immunochemicals 610144121
Encompass
Anti-Mouse IgG DyLight 680 Antibody generated in goat detects reactivity to Mouse IgG. No reaction was observed against Bovine, Chicken, Goat, Guinea Pig, Hamster, Horse, Human, Rabbit, Rat and Sheep Serum Proteins.  This antibody will react with heavy chains of mouse IgG and with light chains of most mouse immunoglobulins. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms. The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Catalog No. 50-105-8406