Learn More
Rockland Immunochemicals Anti-MOUSE IgG (H&L) (GOAT) Antibody DyLight™ 800 Conjugated - 610-145-002
SDP
Supplier:  Rockland Immunochemicals 610145002
Encompass
Anti-Mouse IgG DyLight 800 Antibody generated in goat detects reactivity to Mouse IgG. This antibody will react with heavy chains of Mouse IgG and with light chains of most Mouse immunoglobulins. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition. This product is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms. The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Catalog No. 50-105-8407