Learn More
Rockland Immunochemicals Anti-MOUSE IgG (H&L) (GOAT) Antibody ATTO 488 Conjugated (Min X Bv Ch Gt GP Ham Hs Hu Rb Rt & Sh Serum Proteins) - 610-152-121
SDP
Supplier:  Rockland Immunochemicals 610152121
Encompass
Anti-Mouse IgG (H&L) conjugated by ATTO 488 is designed for STED microscopy, FRET, immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.The emission spectra for this ATTO conjugate matches the principle output wavelengths of most common fluorescence instrumentation.
Catalog No. NC0931390