Learn More
Rockland Immunochemicals Mouse IgG (H&L) Antibody Peroxidase Conjugated 2 mg
SDP
Catalog No. 501943911
Encompass_Preferred
Anti-Mouse IgG (H&L) peroxidase conjugated antibody generated in donkey detects specifically mouse IgG (H&L). Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.
Catalog No. 50-194-3911 Supplier Rockland Immunochemicals Supplier No. 610703002
Anti-Mouse IgG (H&L) peroxidase conjugated antibody generated in donkey detects specifically mouse IgG (H&L). Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition.