Learn More
Rockland Immunochemicals Anti-MOUSE IgG (H&L) (SHEEP) Antibody Peroxidase Conjugated - 610-603-002
SDP
Supplier:  Rockland Immunochemicals 610603002
Encompass
Anti-Mouse IgG whole molecule antibody generated in sheep detects specifically Mouse IgG heavy and light chain. This secondary antibody anti-Mouse is ideal for investigators who routinely perform ELISA, Sandwich ELISA, titration assays, western-blot, immunoprecipitation and more generally immunoassays.
Catalog No. 50-105-8487