Learn More
Cell Signaling Technology Secondary Antibody, Horse Anti-Mouse IgG, HRP-Linked, 5 mL
SDP
Catalog No. NC0976627
Encompass
This affinity-purified horse anti-mouse IgG (heavy and light chain) antibody is conjugated to horseradish peroxidase (HRP) and designed for sensitive chemiluminescent detection. It is rigorously validated for use with primary antibodies to ensure optimal, reproducible performance in western blotting protocols.
  • Conjugated to HRP for chemiluminescent detection
  • Validated for western blotting applications
  • Compatible with diverse chemiluminescent reagents
  • Provides high sensitivity and reliable results
  • Supplied in a stable formulation with glycerol
Catalog No. NC0976627 Supplier Cell Signaling Technology Supplier No. 7076V
This affinity-purified horse anti-mouse IgG (heavy and light chain) antibody is conjugated to horseradish peroxidase (HRP) and designed for sensitive chemiluminescent detection. It is rigorously validated for use with primary antibodies to ensure optimal, reproducible performance in western blotting protocols.
  • Conjugated to HRP for chemiluminescent detection
  • Validated for western blotting applications
  • Compatible with diverse chemiluminescent reagents
  • Provides high sensitivity and reliable results
  • Supplied in a stable formulation with glycerol