Learn More
Seracare Life Sciences Inc ANTI-MOUSE IGM (MU) AB HSA
Supplier:  Seracare Life Sciences Inc 52100184
Encompass_Preferred
Affinity purified polyclonal antibody to Mouse IgM, heavy chain (mu) made in Goat. Product has been cross-adsorbed to human serum to minimize cross-reactivity to human immunoglobulin. Product is in lyophilized form.
Catalog No. 50-668-25