Learn More
Selleck Chemical LLC Anti-mouse IL-4-InVivo
SDP
Supplier:  Selleck Chemical LLC A21095MG
Encompass_Preferred
The 11B11 monoclonal antibody reacts with mouse IL-4 (interleukin-4) which is a multifunctional 14 kDa cytokine The 11B11 monoclonal antibody has been shown to neutralize the bioactivity of natural or recombinant IL-4
Catalog No. 50-234-0835