Learn More
Biolegend Brilliant Violet 785™ anti-mouse Ly-6A/E (Sca-1)
Supplier:  Biolegend 108139
Encompass_Preferred
Brilliant Violet 785™ anti-mouse Ly-6A/E (Sca-1) [D7]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 50 ug
Catalog No. NC1742057