Learn More
STEMCELL Technologies Anti-Mouse Ly-6G Antibody, Clone 1A8, APC, Size: 100 µg
SDP
Supplier:  STEMCELL Technologies 60031AZ
Encompass
Clone 1A8 is a rat monoclonal IgG2a antibody that reacts with mouse Ly-6G, a 21 - 25 kDa GPI-anchored protein, which together with the structurally related Ly-6C protein comprises the granulocyte receptor-1 antigen. Verified for use with flow cytometry assays, use this antibody in applications requiring the detection of Ly-6G. The 1A8 antibody binds specifically to Ly-6G, whereas another commonly used antibody, clone RB6-8C5, binds to both Ly-6G and Ly-6C. It has been reported that the 1A8 antibody detects Ly-6G-expressing granulocytes in peripheral blood, whereas the RB6-8C5 antibody also binds to Ly-6C-expressing lymphocytes, monocytes, and dendritic cells. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ Mouse Neutrophil Enrichment Kit (Catalog #19762). Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5174
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov