Learn More
Selleck Chemical LLC Anti-mouse MHC Class II-InVivo
SDP
Catalog No. 503135667
Encompass_Preferred
Anti-mouse MHC Class II-InVivo reacts with mouse MHC Class II haplotypes I-Ab I-Ad I-Aq I-Ed and I-Ek The antibody does not react with I-Af I-Ak or I-As haplotypes The M5/114 antibody is reported to inhibit I-A-restricted T cell responses
Catalog No. 50-313-5667 Supplier Selleck Chemical LLC Supplier No. A21695MG
Add to Cart
Add to Cart
Anti-mouse MHC Class II-InVivo reacts with mouse MHC Class II haplotypes I-Ab I-Ad I-Aq I-Ed and I-Ek The antibody does not react with I-Af I-Ak or I-As haplotypes The M5/114 antibody is reported to inhibit I-A-restricted T cell responses