Learn More
FUJIFILM Biosciences Anti Mouse Nestin Rat Monoclon
Catalog No. 5000450164
Encompass_Preferred
Nestin is intermediate dilament protein and belongs to a class VI intermediate filament The protein is commonly known neural stem cells and neural progenitor cells marker in developmen of the central nervous system This antibody specifically recognizes mouse Nestin
Catalog No. 50-004-50164 Supplier FUJIFILM Biosciences Supplier No. 01226843
Nestin is intermediate dilament protein and belongs to a class VI intermediate filament The protein is commonly known neural stem cells and neural progenitor cells marker in developmen of the central nervous system This antibody specifically recognizes mouse Nestin