Learn More
STEMCELL Technologies Anti-Mouse OCT4 (OCT3) Antibody, Clone 40, Size: 50 µg
SDP
Supplier:  STEMCELL Technologies 60059
Encompass
Clone 40 is a mouse monoclonal IgG1 antibody that reacts with OCT-3/4, also known as POU5F1, a transcription factor that is essential for the formation of the inner cell mass during mouse development and for the maintenance of undifferentiated mouse and human embryonic stem (ES) and induced pluripotent stem (iPS) cells. Verified for use with flow cytometry, western blotting, and immunofluorescence microscopy assays, use this antibody in applications requiring the detection of OCT4 (OCT3). OCT-3/4 is expressed at high levels in undifferentiated mouse and human ES and iPS cells and in embryonic germ cells (EGCs), and exhibits decreased expression during differentiation. It is commonly used as a marker for the assessment of undifferentiated ES and iPS cells, and EGCs from multiple species. Visit STEMCELL.com for more information and related products.
Catalog No. 50-197-5316
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov