Learn More
FUJIFILM Biosciences Anti Mouse Podoplanin Monoclon
Catalog No. 5000450101
Encompass_Preferred
Podoplanin a type I transmembrane protein having a C-terminal transmembrane region is a glycoprotein with platelet-aggregating activity consisting of 162 and 172 amino acids in humans and mice respectively Podoplanin also called Aggrus or T1a is expressed in lymphatic endothelial cells but not in vascular endothelial cells and therefore has been utilized as a marker specific to lymphatic vessels In brain tumors expression of podoplanin has been known to be increased with increasing WHO grade II III and IV C-type lectin-like receptor 2 CLEC-2 acts as the receptor of podoplanin which contains PLAG2 and PLAG3 domains involved in the binding This antibody is available to detect mouse podoplanin
Catalog No. 50-004-50101 Supplier FUJIFILM Biosciences Supplier No. 01524111
Podoplanin a type I transmembrane protein having a C-terminal transmembrane region is a glycoprotein with platelet-aggregating activity consisting of 162 and 172 amino acids in humans and mice respectively Podoplanin also called Aggrus or T1a is expressed in lymphatic endothelial cells but not in vascular endothelial cells and therefore has been utilized as a marker specific to lymphatic vessels In brain tumors expression of podoplanin has been known to be increased with increasing WHO grade II III and IV C-type lectin-like receptor 2 CLEC-2 acts as the receptor of podoplanin which contains PLAG2 and PLAG3 domains involved in the binding This antibody is available to detect mouse podoplanin