Learn More
Medchemexpress LLC Anti-Mouse Rat Human 1mg | 1mg
SDP
Catalog No. 5000325716
Encompass_Preferred
Anti-Mouse/Rat/Human CCL2/MCP-1 Antibody (2H5) is an anti-mouse/rat/human CCL2/MCP-1 IgG monoclonal antibody Anti-Mouse/Rat/Human CCL2/MCP-1 Antibody (2H5) can reverse the immunosuppressive microenvironment by blocking the CCL2 signaling pathway Anti-Mouse/Rat/Human CCL2/MCP-1 Antibody (2H5) can reduce the secretion of IFN- and the infiltration of macrophages CCL2/MCP-1 Antibody (2H5) can reduce the amount of HIV virus by increasing the proportion of T cells Anti-Mouse/Rat/Human CCL2/MCP-1 Antibody (2H5) can be used for researches on inflammation conditions virus infection and cancer such as inflammatory bowel disease (IBD) hepatocellular carcinoma (HCC) and HIV[1][2][3][4][5]
Catalog No. 50-003-25716 Supplier Medchemexpress LLC Supplier No. HYP9902971MG
Anti-Mouse/Rat/Human CCL2/MCP-1 Antibody (2H5) is an anti-mouse/rat/human CCL2/MCP-1 IgG monoclonal antibody Anti-Mouse/Rat/Human CCL2/MCP-1 Antibody (2H5) can reverse the immunosuppressive microenvironment by blocking the CCL2 signaling pathway Anti-Mouse/Rat/Human CCL2/MCP-1 Antibody (2H5) can reduce the secretion of IFN- and the infiltration of macrophages CCL2/MCP-1 Antibody (2H5) can reduce the amount of HIV virus by increasing the proportion of T cells Anti-Mouse/Rat/Human CCL2/MCP-1 Antibody (2H5) can be used for researches on inflammation conditions virus infection and cancer such as inflammatory bowel disease (IBD) hepatocellular carcinoma (HCC) and HIV[1][2][3][4][5]