Learn More
Medchemexpress LLC Anti-mouse/rat/human cd47/iap antibody (miap410) | 99.6% | 150 kDa | 5 MG
SDP
Catalog No. 5000393447
Encompass_Preferred
The Anti-Mouse/Rat/Human CD47/IAP Antibody (MIAP410) is an IgG1 monoclonal antibody designed to effectively block CD47 signaling and enhance macrophage phagocytic function. It can increase immune cell infiltration and may be involved in wound healing. This antibody is suitable for research into cancer and inflammation conditions, including breast cancer and intestinal mucosal repair.
  • Effectively blocks CD47 signaling.
  • Enhances macrophage phagocytic function.
  • Increases the infiltration of immune cells.
  • Suitable for cancer and inflammation research.
  • Inhibits epithelial cell migration and wound closure.
  • Inhibits mucosal wound healing.
  • Inhibits tumor growth.
  • Does not cause significant toxicity.
Catalog No. 50-003-93447 Supplier Medchemexpress LLC Supplier No. 5MGHYP990132
The Anti-Mouse/Rat/Human CD47/IAP Antibody (MIAP410) is an IgG1 monoclonal antibody designed to effectively block CD47 signaling and enhance macrophage phagocytic function. It can increase immune cell infiltration and may be involved in wound healing. This antibody is suitable for research into cancer and inflammation conditions, including breast cancer and intestinal mucosal repair.
  • Effectively blocks CD47 signaling.
  • Enhances macrophage phagocytic function.
  • Increases the infiltration of immune cells.
  • Suitable for cancer and inflammation research.
  • Inhibits epithelial cell migration and wound closure.
  • Inhibits mucosal wound healing.
  • Inhibits tumor growth.
  • Does not cause significant toxicity.