Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Anti-Mouse/Rat/Human LRP1/CD91 Antibody (11H4) | ≥95.0% | 150 kDa | 5 MG
SDP

Catalog No. 5000393464
Encompass_Preferred

Anti-Mouse/Rat/Human LRP1/CD91 Antibody (11H4) is a mouse-derived IgG1 κ type antibody inhibitor, targeting mouse/rat/human LRP1/CD91. It binds to the β chain of LRP1. This antibody affects intracellular signaling, lipid homeostasis, and clearance of apoptotic cells by blocking LRP1. It can be used for western blot (WB), immunofluorescence (IF), and other detections. This product is for research use only.

  • Isotype: Mouse IgG1 kappa
  • Species reactivity: Human/mouse/rat
  • Target: LRP1/CD91
  • Application: ELISA, FACS, functional assay, research in vivo
  • Conjugated: Unconjugated
  • Appearance: Liquid, colorless to light yellow
  • Usage: For research use only
  • Can be used for western blot (WB), immunofluorescence (IF), and other detections
  • Affects intracellular signaling, lipid homeostasis, and clearance of apoptotic cells by blocking LRP1
  • Binds to the β chain of LRP1

Catalog No. 50-003-93464 Supplier Medchemexpress LLC Supplier No. 5MGHYP990202
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Anti-Mouse/Rat/Human LRP1/CD91 Antibody (11H4) is a mouse-derived IgG1 κ type antibody inhibitor, targeting mouse/rat/human LRP1/CD91. It binds to the β chain of LRP1. This antibody affects intracellular signaling, lipid homeostasis, and clearance of apoptotic cells by blocking LRP1. It can be used for western blot (WB), immunofluorescence (IF), and other detections. This product is for research use only.

  • Isotype: Mouse IgG1 kappa
  • Species reactivity: Human/mouse/rat
  • Target: LRP1/CD91
  • Application: ELISA, FACS, functional assay, research in vivo
  • Conjugated: Unconjugated
  • Appearance: Liquid, colorless to light yellow
  • Usage: For research use only
  • Can be used for western blot (WB), immunofluorescence (IF), and other detections
  • Affects intracellular signaling, lipid homeostasis, and clearance of apoptotic cells by blocking LRP1
  • Binds to the β chain of LRP1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.