Learn More
Medchemexpress LLC Anti-Mouse Rat IL-17 5mg | 5mg
SDP
Catalog No. 5000325175
Encompass_Preferred
Anti-Mouse/Rat IL-17A Antibody (17F3) is a mouse-derived IgG1 type antibody inhibitor targeting to mouse and rat IL-17A Anti-Mouse IL-17A Antibody (17F3) can neutralize IL-17 Anti-Mouse/Rat IL-17A Antibody (17F3) can be used for the researches of infection immunology and metabolic disease such as M intracellulare infection and diabetes[1][2][3]
Catalog No. 50-003-25175 Supplier Medchemexpress LLC Supplier No. HYP9902225MG
Anti-Mouse/Rat IL-17A Antibody (17F3) is a mouse-derived IgG1 type antibody inhibitor targeting to mouse and rat IL-17A Anti-Mouse IL-17A Antibody (17F3) can neutralize IL-17 Anti-Mouse/Rat IL-17A Antibody (17F3) can be used for the researches of infection immunology and metabolic disease such as M intracellulare infection and diabetes[1][2][3]