Learn More
Selleck Chemical LLC Anti-mouse/rat IL-b-InVivo
SDP
Catalog No. 503138859
Encompass_Preferred
Anti-mouse/rat IL-1 -InVivo reacts with precursor and mature secreted forms of mouse and rat IL-1 a 17 kDa pro-inflammatory cytokine produced primarily by monocytes
Catalog No. 50-313-8859 Supplier Selleck Chemical LLC Supplier No. A21421MG
Anti-mouse/rat IL-1 -InVivo reacts with precursor and mature secreted forms of mouse and rat IL-1 a 17 kDa pro-inflammatory cytokine produced primarily by monocytes