Learn More
Selleck Chemical LLC Anti-mouse Ter-119-InVivo-1MG
SDP
Catalog No. 503138700
Encompass_Preferred
Anti-mouse Ter-119-InVivo reacts with mouse Ter-119 a 52 kDa glycophorin A-associated protein that is expressed by erythroid cells from the early proerythroblast stage to mature erythrocytes Anti-mouse Ter-119-InVivo is commonly used for identifying erythrocytes and cells in the erythroid lineage
Catalog No. 50-313-8700 Supplier Selleck Chemical LLC Supplier No. A21411MG
Anti-mouse Ter-119-InVivo reacts with mouse Ter-119 a 52 kDa glycophorin A-associated protein that is expressed by erythroid cells from the early proerythroblast stage to mature erythrocytes Anti-mouse Ter-119-InVivo is commonly used for identifying erythrocytes and cells in the erythroid lineage