Learn More
Selleck Chemical LLC Anti-mouse Ter-119-InVivo
SDP
Supplier:  Selleck Chemical LLC A21415MG 5PC
Encompass_Preferred
Anti-mouse Ter-119-InVivo reacts with mouse Ter-119 a 52 kDa glycophorin A-associated protein that is expressed by erythroid cells from the early proerythroblast stage to mature erythrocytes Anti-mouse Ter-119-InVivo is commonly used for identifying erythrocytes and cells in the erythroid lineage
Catalog No. 50-313-8011