Learn More
Medchemexpress LLC Anti-Mouse Thy1 CD90 5mg | 5mg
SDP
Catalog No. 5000325149
Encompass_Preferred
Anti-Mouse Thy1/CD90 Antibody (M5/49 4 1) is an anti-mouse Thy1/CD90 IgG2a monoclonal antibody Anti-Mouse Thy1/CD90 Antibody (M5/49 4 1) can clear T cells from the spleen and bone marrow Anti-Mouse Thy1/CD90 Antibody (M5/49 4 1) can be used for researches on immunology and inflammation conditions such as colitis[1][2][3]
Catalog No. 50-003-25149 Supplier Medchemexpress LLC Supplier No. HYP9901575MG
Anti-Mouse Thy1/CD90 Antibody (M5/49 4 1) is an anti-mouse Thy1/CD90 IgG2a monoclonal antibody Anti-Mouse Thy1/CD90 Antibody (M5/49 4 1) can clear T cells from the spleen and bone marrow Anti-Mouse Thy1/CD90 Antibody (M5/49 4 1) can be used for researches on immunology and inflammation conditions such as colitis[1][2][3]