Learn More
Bio X Cell InVivoMAb anti-mouse VEGFR-2, Clone DC101, 100 MG
Supplier: Bio X Cell BE0060R100MG
The DC101 monoclonal antibody targets mouse VEGFR-2 (vascular endothelial growth factor receptor 2), which is a tyrosine protein kinase essential for vascular development and permeability. This antibody inhibits VEGFR-2 signaling in vivo, making it suitable for research applications.
- Isotype: Rat IgG1, κ
- Conjugation: Unconjugated
- Immunogen: Mouse VEGFR-2-SEAPs soluble receptor
- Applications: In vivo/in vitro blocking of VEGF/VEGFR-2 signaling, Western blot
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtration
- Purification: Protein G, animal-free facility
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.