Learn More
Medchemexpress LLC Anti-Mouse VEGFR-2 A 1mg | 1mg
SDP
Catalog No. 5000324666
Encompass_Preferred
Anti-Mouse VEGFR-2 Antibody (DC101) is a rat anti-mouse VEGFR2 monoclonal antibody Anti-Mouse VEGFR-2 Antibody (DC101) inhibits tumor angiogenesis by blocking the binding of VEGF and VEGFR2 Anti-Mouse VEGFR-2 Antibody (DC101) promotes immune cell infiltration and induces tumor cell apoptosis Anti-Mouse VEGFR-2 Antibody (DC101) can be used for researches on various types of cancer such as melanoma lung cancer and breast cancer [1][2][3][4][5][6]
Catalog No. 50-003-24666 Supplier Medchemexpress LLC Supplier No. HYP9901061MG
Anti-Mouse VEGFR-2 Antibody (DC101) is a rat anti-mouse VEGFR2 monoclonal antibody Anti-Mouse VEGFR-2 Antibody (DC101) inhibits tumor angiogenesis by blocking the binding of VEGF and VEGFR2 Anti-Mouse VEGFR-2 Antibody (DC101) promotes immune cell infiltration and induces tumor cell apoptosis Anti-Mouse VEGFR-2 Antibody (DC101) can be used for researches on various types of cancer such as melanoma lung cancer and breast cancer [1][2][3][4][5][6]