Learn More
Seracare Life Sciences Inc ANTI-MS A+G+M (H+L) AB FITC
Supplier:  Seracare Life Sciences Inc 52300308
Encompass_Preferred
Affinity purified polyclonal secondary antibody against Mouse IgA, IgG, and IgM, both heavy and light chains (whole IgA, IgM, and IgG) made in Goat and labeled with fluorescein isothiocyanate (FITC).
Catalog No. 50-668-94