Learn More
Leinco Technologies Inc Anti-Mouse M-CSFR (CSF-1R, M-CSFR, c-fms) In Vivo Platinum; Clone AFS98; Isotype: Rat IgG2a κ
SDP
Supplier:  Leinco Technologies Inc C2268100 MG
Encompass_Preferred
Specificity: Clone AFS98 recognizes an epitope on mouse M-CSFR. Antigen Distribution: M-CSFR is expressed on monocytes/macrophages, peritoneal exudate cells, plasmacytoid cells, dendritic cells, and osteoclasts. Background: M-CSFR is a 145 kD protein, is part of the Ig superfamily, and triggers growth and development of mononuclear phagocytes and placental trophoblasts. M-CSF and its receptor present as candidate therapeutic targets in states of inflammatory bone erosion. Applications: (WB, B, FA) When M-CSF ligand specifically binds to the M-CSF receptor on osteoclasts, it induces receptor dimerization, activation and autophosphorylation of cytoplasmic tyrosine residues. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-5904