Learn More
Leinco Technologies Inc Anti-Mouse CD11a (LFA-1α) In Vivo GOLD; Clone I21/7; Isotype: Rat IgG2a
SDP
Supplier:  Leinco Technologies Inc C37625 MG
Encompass_Preferred
Specificity: Clone I21/7 recognizes mouse CD11a. Antigen Distribution: CD11a is present on thymocytes, blood lymphocytes, bone marrow cells and certain lymphoma and macrophage-like cell lines. Background: CD11a is a180 kD glycoprotein and is the α subunit of LFA-1. LFA-1 is a member of a family of heterodimeric receptors which mediate cellular adhesion reactions through interaction with ICAM-1 and ICAM-2, including participation in the immunological synapses between CD8+ T lymphocytes and antigen-presenting cells. Applications: (IP, WB, B, FA) Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6103