Learn More
Leinco Technologies Inc Anti-Mouse CD11c (CR4, p150, ITGAX) In Vivo GOLD; Clone N418; Isotype: Armenian Hamster IgG
SDP
Supplier:  Leinco Technologies Inc C21195.0 MG
Encompass_Preferred
Specificity: Clone N418 recognizes an epitope on mouse CD11C. Antigen Distribution: The antigen is primarily expressed on dendritic cells, NK cells, a subset of intestinal intraepithelial lymphocytes (IEL), and some activated T cells. Background: CD11c is a 150 kD glycoprotein. Applications: (WB, IP, FC) This product is validated to work with CyTOF. Other reported applications are immunohistochemical staining of acetone-fixed frozen sections and immunofluorescence. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-5879