Learn More
Leinco Technologies Inc Anti-Mouse CD120α (CD120a, TNFR1, TNF R Type I/p55) In Vivo GOLD; Clone 55R-593; Isotype: Armenian Hamster IgG
SDP
Supplier:  Leinco Technologies Inc T2451.0 MG
Encompass_Preferred
Specificity: Clone 55R-593 recognizes mouse CD120α. Antigen Distribution: CD120α is expressed in various cell types at low levels. Background: CD120α is a 55kD Type I transmembrane protein receptor that binds both TNF-α and TNF-β (LT-α). In association with TRADD and RIP, the receptor crosslinking induced by TNF-α or TNF-β trimers is vital for signal transduction, leading to apoptosis, NF-kB activation, increased expression of proinflammatory genes, tumor necrosis, and cell differentiation depending on cell type and differentiation state. Applications: (IP, WB) Clone 55R-593 has been reported to block ligand-induced receptor signaling. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-6645