Learn More
Leinco Technologies Inc Anti-Mouse CD155 (PVR (poliovirus receptor) homolog) In Vivo GOLD; Clone: 4.24 (4.24.1); Isotype: Rat IgG2a κ
SDP
Supplier:  Leinco Technologies Inc C283325 MG
Encompass_Preferred
Specificity: Clone 4.24.1 recognizes an epitope on mouse CD155. Antigen Distribution: CD155 is expressed at cell junctions on the primary vascular endothelial cells and is highly expressed on DP thymocytes. Background: CD155 is a member of a subfamily of immunoglobulin-like adhesion receptors. Apart from its existence on epithelial cells, this antigen is expressed on a variety of cells of hematopoietic origin. Applications: (WB, FA) The engagement of CD155 with CD226 and CD96 induces cytotoxicity of NK cells and CTL. Leinco’s In vivo GOLDTM and In vivo PLATINUMTM functional antibodies are manufactured in an animal free facility using only In vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates.
Catalog No. 50-190-5942